Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Silver protein

Supplier: BEANTOWN CHEMICAL - BTC

121335-100G 121335-25G 121335-5G
BT121335-100GEA 290 USD InStock
BT121335-100G BT121335-25G BT121335-5G
Silver protein
Silver protein
RTECS: UK9510000
Form: Powder
Sensitivity: Hygroscopic, Light sensitive
Merck Number: 14,8527


Formula: Ag (with protein)
MDL Number: MFCD00132268
CAS Number: 9015-51-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty