Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zeolite naturally occurring mineral

Supplier: BEANTOWN CHEMICAL - BTC

121420-1PC 121420-5PC
BT121420-1PCEA 88 USD InStock
BT121420-1PC BT121420-5PC
Zeolite naturally occurring mineral
Zeolite
CAS: 68989-22-0; MDL No: MFCD00132601 Pieces

  • Pieces


Formula: Na₂O·Al₂O₃·xSiO₂·yH₂O
MDL Number: MFCD00132601
CAS Number: 1318-02-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty