Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3'-Chloro-2'-fluoroacetophenone ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

121795-1G 121795-25G 121795-5G
BT121795-1GEA 49 USD InStock
BT121795-1G BT121795-25G BT121795-5G
3'-Chloro-2'-fluoroacetophenone ≥97%
3'-Chloro-2'-fluoroacetophenone
CAS: 161957-59-1; MDL No: MFCD00106165 Liquid; Molecular Formula: C8H6ClFO; MW: 172.59 Refractive Index: 1.532

  • Refractive index: 1.532
  • Liquid


Formula: C₈H₆ClFO
MW: 172.59
MDL Number: MFCD00106165
CAS Number: 161957-59-1
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty