Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Itaconic acid ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

122075-1KG 122075-250G 122075-5KG
BT122075-1KGEA 35 USD InStock
BT122075-1KG BT122075-250G BT122075-5KG
Itaconic acid ≥99%
Itaconic acid
CAS: 97-65-4; EC No: 202-599-6; MDL No: MFCD00004260 Solid; Molecular Formula: C5H6O4; MW: 130.10 Melting Point: 165-168° Density (g/mL): 1.573 Hygroscopic

  • Hygroscopic
  • Solid


Formula: C₅H₆O₄
MW: 130.1 g/mol
Boiling Pt: 381
Melting Pt: 165…168 °C
Density: 1.573 g/mL
MDL Number: MFCD00004260
CAS Number: 97-65-4
EINECS: 202-599-6
Merck Index: 13,05264
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty