Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1'-Thiocarbonyldiimidazole ≥90%, Technical Grade

Supplier: BEANTOWN CHEMICAL - BTC

122495-10G 122495-250G 122495-50G
BT122495-10GEA 95 USD InStock
BT122495-10G BT122495-250G BT122495-50G
1,1'-Thiocarbonyldiimidazole ≥90%, Technical Grade
1,1'-Thiocarbonyldiimidazole
CAS: 6160-65-2; EC No: 228-183-4; MDL No: MFCD00005289 Powder; Molecular Formula: C7H6N4S; MW: 178.21 Melting Point: 98-102° Moisture Sensitive

  • Moisture Sensitive
  • Powder


Formula: C₇H₆N₄S
MW: 178.22 g/mol
Melting Pt: 98…102 °C
MDL Number: MFCD00005289
CAS Number: 6160-65-2
EINECS: 228-183-4
Order Now
Specification Test Results
Assay 90%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty