Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl alcohol ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

122715-1KG 122715-250G
BT122715-1KGEA 78.71 USD InStock
BT122715-1KG BT122715-250G
Benzyl alcohol ≥99%
Benzyl alcohol
CAS: 100-51-6; EC No: 202-859-9; MDL No: MFCD00004599; RTECS: DN3150000 Liquid; Molecular Formula: C7H8O ; MW: 108.14 Melting Point: -16°; Boiling Point: 203-205°; Flash point: 96°C (205°F) Density (g/mL): 1.045; Refractive Index: 1.539

  • RTECS: DN3150000
  • Refractive index: 1.539
  • Liquid


Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 205
Melting Pt: -15.3
Density: 1.045 g/mL
Flash Pt: 96 °C (205 °F)
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=fc6a85fbdb3ccffa1f6db5eed6a9883a84e92c6b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty