Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Iron(III) hexacyanoferrate(II)

Supplier: BEANTOWN CHEMICAL - BTC

123255-100G 123255-25G
BT123255-100GEA 165 USD InStock
BT123255-100G BT123255-25G
Iron(III) hexacyanoferrate(II)
Iron(III) hexacyanoferrate(II)
CAS: 14038-43-8; EC No: 237-875-5; MDL No: MFCD00135663 Powder; Linear Formula: Fe4[Fe(CN)6]3; Molecular Formula: C18Fe7N18; MW: 859.25 Hygroscopic

  • Hygroscopic
  • Powder


Formula: C₁₈Fe₇N₁₈
MW: 859.24 g/mol
MDL Number: MFCD00135663
CAS Number: 14038-43-8
EINECS: 237-875-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty