Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-Bromononanoic acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

123695-1G 123695-5G
BT123695-1GEA 141 USD InStock
BT123695-1G BT123695-5G
9-Bromononanoic acid ≥98%
9-Bromononanoic acid
CAS: 41059-02-3; MDL No: MFCD00046564 Solid; Linear Formula: Br(CH2)8COOH; MW: 237.13 Melting Point: 39°; Boiling Point: 317°

  • Solid


Formula: C₉H₁₇BrO₂
Boiling Pt: 317 °C
Melting Pt: 39 °C
MDL Number: MFCD00046564
CAS Number: 41059-02-3
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty