Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-D-Pro-OH ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

124295-1G 124295-5G
BT124295-1GEA 62 USD InStock
BT124295-1G BT124295-5G
Boc-D-Pro-OH ≥98%
Boc-D-Pro-OH
CAS: 37784-17-1; MDL No: MFCD00063226 Powder; Molecular Formula: C10H17NO4; MW: 215.25 Melting Point: 134-137° Optical Rotation: [α]22/D +60°, c = 2 in acetic acid

  • Optical rotation: [α]22/D +60°, c = 2 in acetic acid
  • Powder


Formula: C₁₀H₁₇NO₄
Melting Pt: 134…137 °C
MDL Number: MFCD00063226
CAS Number: 37784-17-1
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty