Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Silver ≥99.999%, shot 1-3 mm

Supplier: BEANTOWN CHEMICAL - BTC

124905-10G 124905-250G 124905-50G
BT124905-10GEA 113 USD InStock
BT124905-10G BT124905-250G BT124905-50G
Silver ≥99.999%, shot 1-3 mm
Silver
CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397; RTECS: VW3500000 Solid; Molecular Formula: Ag; MW: 107.87 Melting Point: 960°; Boiling Point: 2212° Density (g/mL): 10.49

  • RTECS: VW3500000
  • Solid


Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.49 g/mL
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647
Order Now
Specification Test Results
Assay 99.999%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty