Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tin ≥99.995% (trace metals basis), foil, Thickness: 0.25 mm

Supplier: BEANTOWN CHEMICAL - BTC

125000-100X100 125000-100X500 125000-50X50MM
BT125000-100X1EA 170 USD InStock
BT125000-100X1 BT125000-100X5 BT125000-50X50
Tin ≥99.995% (trace metals basis), foil, Thickness: 0.25 mm
Tin
CAS: 7440-31-5; EC No: 231-141-8; MDL No: MFCD00133862 Solid; Molecular Formula: Sn; MW: 118.71 Melting Point: 231.9°; Boiling Point: 2270° Density (g/mL): 7.31

  • Solid


Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270 °C
Melting Pt: 231.9 °C
Density: 7.31 g/mL
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587
Order Now
Specification Test Results
Assay 99.995%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty