Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium ferulate

Supplier: BEANTOWN CHEMICAL - BTC

125135-1G 125135-5G
BT125135-1GEA 197 USD InStock
BT125135-1G BT125135-5G
Sodium ferulate
Sodium ferulate
CAS: 24276-84-4; MDL No: MFCD00075727 Solid; Molecular Formula: C10H9NaO4; MW: 216.17

  • Solid


Formula: C₁₀H₉NaO₄
MW: 216.168 g/mol
MDL Number: MFCD00075727
CAS Number: 24276-84-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty