Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Kerosene

Supplier: BEANTOWN CHEMICAL - BTC

125280-2.5L 125280-500ML
BT125280-2.5LEA 60.6 USD InStock
BT125280-2.5L BT125280-500ML
Kerosene
Kerosene - CAS 64742-47-8
CAS: 64742-47-8; EC No: 265-149-8; MDL No: MFCD00135561; RTECS: OA5500000 Liquid Boiling Point: 175-325°; Flash point: 76°C (169°F) Density (g/mL): 0.8

  • RTECS: OA5500000
  • Liquid


Boiling Pt: 175…325 °C
Density: 0.8 g/mL
Flash Pt: 76 °C (169 °F)
CAS Number: 64742-47-8
EINECS: 265-149-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty