Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Iodophenol ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

125295-10G 125295-250G 125295-50G
BT125295-10GEA 45 USD InStock
BT125295-10G BT125295-250G BT125295-50G
3-Iodophenol ≥98%
3-Iodophenol
CAS: 626-02-8; EC No: 210-923-2; MDL No: MFCD00002261; RTECS: SL5490000 Solid; Molecular Formula: C6H5IO; MW: 220.01 Melting Point: 42-44°; Flash point: 110°C (230°F) Light Sensitive

  • RTECS: SL5490000
  • Light Sensitive
  • Solid


Formula: C₆H₅IO
Melting Pt: 42…44 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00002261
CAS Number: 626-02-8
EINECS: 210-923-2
UN: 3335
ADR: 9,
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty