Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perfluoro-1,2-dimethylcyclobutane ≥97% main impurity is 1,3-isomer

Supplier: BEANTOWN CHEMICAL - BTC

125475-100G 125475-25G
BT125475-100GEA 201 USD InStock
BT125475-100G BT125475-25G
Perfluoro-1,2-dimethylcyclobutane ≥97% main impurity is 1,3-isomer
Perfluoro-1,2-dimethylcyclobutane
CAS: 2994-71-0; EC No: 221-065-3; MDL No: MFCD00013737 Liquid; Molecular Formula: C6F12; MW: 300.05 Melting Point: -32°; Boiling Point: 45° Density (g/mL): 1.62; Refractive Index: 1.254

  • Refractive index: 1.254
  • Liquid


Formula: C₆F₁₂
Boiling Pt: 45 °C
Melting Pt: –32 °C
Density: 1.62 g/mL
MDL Number: MFCD00013737
CAS Number: 2994-71-0
EINECS: 221-065-3
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty