Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Itaconic anhydride ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

126290-100G 126290-25G 126290-500G
BT126290-100GEA 100 USD InStock
BT126290-100G BT126290-25G BT126290-500G
Itaconic anhydride ≥97%
Itaconic anhydride
CAS: 2170-03-8; EC No: 218-518-2; MDL No: MFCD00005530 Powder; Molecular Formula: C5H4O3; MW: 112.09 Melting Point: 66-68°; Boiling Point: 114-115°/12 mmHg Moisture Sensitive

  • Moisture Sensitive
  • Powder


Formula: C₅H₄O₃
MW: 112.08 g/mol
Boiling Pt: 239
Melting Pt: 66…68 °C
MDL Number: MFCD00005530
CAS Number: 2170-03-8
EINECS: 218-518-2
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty