Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Perfluorocyclohexyl)methyl acrylate ≥96% stabilised

Supplier: BEANTOWN CHEMICAL - BTC

126420-1G 126420-25G 126420-5G
BT126420-1GEA 33 USD InStock
BT126420-1G BT126420-25G BT126420-5G
(Perfluorocyclohexyl)methyl acrylate ≥96% stabilised
(Perfluorocyclohexyl)methyl acrylate
CAS: 40677-94-9; MDL No: MFCD00080614 Liquid; Molecular Formula: C10H5F11O2; MW: 366.13 Air Sensitive

  • Air Sensitive
  • Liquid


Formula: C₁₀H₅F₁₁O₂
MW: 366.13 g/mol
MDL Number: MFCD00080614
CAS Number: 40677-94-9
Order Now
Specification Test Results
Assay 96%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty