Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Gold ≥99.95%, soluble APS 1,5-3,0 µm

Supplier: BEANTOWN CHEMICAL - BTC

130420-1G 130420-5G
BT130420-1GEA 211 USD InStock
BT130420-1G BT130420-5G
Gold ≥99.95%, soluble APS 1,5-3,0 µm
Gold
CAS: 7440-57-5; EC No: 231-165-9; MDL No: MFCD00003436 Solid; Molecular Formula: Au; MW: 196.97

  • Solid


Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526
Order Now
Specification Test Results
Assay 99.95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty