Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc citrate dihydrate

Supplier: BEANTOWN CHEMICAL - BTC

135315-1KG 135315-250G
BT135315-1KGEA 121 USD InStock
BT135315-1KG BT135315-250G
Zinc citrate dihydrate
Zinc citrate dihydrate
CAS: 5990-32-9; EC No: 208-901-2; MDL No: MFCD00150587; RTECS: GE8360000 Powder; Linear Formula: (C6H5O7)2Zn3·2H2O; MW: 610.40 Melting Point: 334° (decomposes)

  • RTECS: GE8360000
  • Powder


Formula: C₁₂H₁₄O₁₆Zn₃
MW: 610.368 g/mol
Melting Pt: 334 °C (dec.)
MDL Number: MFCD00150587
CAS Number: 5990-32-9
EINECS: 208-901-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty