Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminonicotinic acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

135405-100G 135405-25G 135405-5G
BT135405-100GEA 610 USD InStock
BT135405-100G BT135405-25G BT135405-5G
2-Aminonicotinic acid ≥98%
2-Aminonicotinic acid
CAS: 5345-47-1; EC No: 226-296-3; MDL No: MFCD00006318 Crystalline/Powder; Molecular Formula: C6H6N2O2 ; MW: 138.12 Melting Point: 295-297° (decomposes)

  • Crystalline/Powder


Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: 297 (dec.)
MDL Number: MFCD00006318
CAS Number: 5345-47-1
EINECS: 226-296-3
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty