Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Hexadecene ≥94%

Supplier: BEANTOWN CHEMICAL - BTC

144660-1L 144660-250ML
BT144660-1LEA 128 USD InStock
BT144660-1L BT144660-250ML
1-Hexadecene ≥94%
1-Hexadecene
CAS: 629-73-2; EC No: 211-105-8; MDL No: MFCD00008991; RTECS: MM0479000 Liquid; Linear Formula: CH3(CH2)13CH=CH2; Molecular Formula: C16H31; MW: 224.43 Melting Point: 3-5°; Boiling Point: 274°; Flash point: 132°C (270°F) Density (g/mL): 0.783; Refractive Index: 1.441

  • RTECS: MM0479000
  • Refractive index: 1.441
  • Liquid


Formula: C₁₆H₃₂
MW: 224.43 g/mol
Boiling Pt: 285
Melting Pt: 4
Density: 0.783 g/mL
Flash Pt: 132 °C (270 °F)
MDL Number: MFCD00008991
CAS Number: 629-73-2
EINECS: 211-105-8
Order Now
Specification Test Results
Assay 94%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty