127150-100X150
127150-150X300
127150-300X600
BT127150-100X1EA
54
USD
InStock
BT127150-100X1
BT127150-150X3
BT127150-300X6
Molybdenum ≥99.95% (trace metals basis), foil, Thickness 0.1 mm
Molybdenum
CAS: 7439-98-7; EC No: 231-107-2; MDL No: MFCD00003465; RTECS: QA4680000
Solid; Molecular Formula: Mo; MW: 95.94
Melting Point: 2617°; Boiling Point: 4612°
Density (g/mL): 10.3