Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl cinnamate ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

127305-100G 127305-2.5KG 127305-500G
BT127305-100GEA 26 USD InStock
BT127305-100G BT127305-2.5KG BT127305-500G
Methyl cinnamate ≥99%
Methyl cinnamate
CAS: 103-26-4; EC No: 203-093-8; MDL No: MFCD00008458; RTECS: GE0190000 Crystalline; Linear Formula: C6H5CH=CHCOOCH3; Molecular Formula: C10H10O2; MW: 162.19 Melting Point: 33-38°; Boiling Point: 260-262°; Flash point: 110°C (230°F) Density (g/mL): 1.092

  • RTECS: GE0190000
  • Crystalline


Formula: C₁₀H₁₀O₂
Boiling Pt: 260…262 °C
Melting Pt: 33…38 °C
Density: 1.092 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00008458
CAS Number: 103-26-4
EINECS: 203-093-8
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty