Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl sulphoxide ≥99.9% ACS

Supplier: BEANTOWN CHEMICAL - BTC

127445-1L 127445-4L 127445-500ML
BT127445-1LEA 106.78 USD InStock
BT127445-1L BT127445-4L BT127445-500ML
Dimethyl sulphoxide ≥99.9% ACS
Dimethyl sulphoxide
CAS: 67-68-5; EC No: 200-664-3; MDL No: MFCD00002089; RTECS: PV6210000 Liquid; Linear Formula: (CH3)2SO ; MW: 78.13 Melting Point: 16-19°; Boiling Point: 189°; Flash point: 87°C (189°F) Density (g/mL): 1.101; Refractive Index: 1.479 Hygroscopic

  • RTECS: PV6210000
  • Refractive index: 1.479
  • Hygroscopic
  • Liquid


Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189
Melting Pt: 18.5
Density: 1.101 g/mL
Flash Pt: 87 °C (189 °F)
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=d0727d3220885f6adab5f099d9fd3b4de38d5710&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99.9% min.
Residue on evaporation ≤0.008%
Water ≤0.07%
Acid ≤0.001meq/g
Appearance Clear, colorless, free of sediment

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty