Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethallyl carbonate ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

127930-25G 127930-5G
BT127930-25GEA 153 USD InStock
BT127930-25G BT127930-5G
Dimethallyl carbonate ≥97%
Dimethallyl carbonate
CAS: 64057-79-0; EC No: 264-645-1; MDL No: MFCD00008598; RTECS: FF8750000 Liquid; Molecular Formula: C11H18O3; MW: 170.21 Boiling Point: 199-202°

  • RTECS: FF8750000
  • Liquid


Formula: C₉H₁₄O₃
MW: 170.208 g/mol
Boiling Pt: 199…202 °C
MDL Number: MFCD00008598
CAS Number: 64057-79-0
EINECS: 264-645-1
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty