128385-150MM
128385-50MM
BT128385-150MMEA
288
USD
InStock
BT128385-150MM
BT128385-50MM
Cobalt ≥99.95% (trace metals basis), rods Ø 5 mm
Cobalt
CAS: 7440-48-4; EC No: 231-158-0; MDL No: MFCD00010935; RTECS: GF8750000
Solid; Molecular Formula: Co; MW: 58.93
Melting Point: 1495°; Boiling Point: 2900°
Density (g/mL): 8.92