Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl fumarate ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

128695-100G 128695-250G 128695-25G
BT128695-100GEA 58 USD InStock
BT128695-100G BT128695-250G BT128695-25G
Dimethyl fumarate ≥99%
Dimethyl fumarate
CAS: 624-49-7; EC No: 210-849-0; MDL No: MFCD00064438; RTECS: EM6125000 Powder; Linear Formula: CH3OCOCH=CHCOOCH3; Molecular Formula: C6H8O4 ; MW: 144.13 Melting Point: 102-106°; Boiling Point: 192-193°

  • RTECS: EM6125000
  • Powder


Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 192
Melting Pt: 102
MDL Number: MFCD00064438
CAS Number: 624-49-7
EINECS: 210-849-0
Merck Index: 13,04308
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty