Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(3-Methoxyphenyl)propionic acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

129115-25G 129115-5G
BT129115-25GEA 160 USD InStock
BT129115-25G BT129115-5G
3-(3-Methoxyphenyl)propionic acid ≥98%
3-(3-Methoxyphenyl)propionic acid
CAS: 10516-71-9; EC No: 234-049-6; MDL No: MFCD00014027 Crystalline; Molecular Formula: C10H12O3; MW: 180.20 Melting Point: 43-45°

  • Crystalline


Formula: C₁₀H₁₂O₃
Melting Pt: 43…45 °C
MDL Number: MFCD00014027
CAS Number: 10516-71-9
EINECS: 234-049-6
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty