Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Gold ≥99.995% (trace metals basis), foil, Thickness: 0.25 mm

Supplier: BEANTOWN CHEMICAL - BTC

129240-25X25MM 129240-50X50MM
BT129240-25X25EA 528 USD InStock
BT129240-25X25 BT129240-50X50
Gold ≥99.995% (trace metals basis), foil, Thickness: 0.25 mm
Gold
CAS: 7440-57-5; EC No: 231-165-9; MDL No: MFCD00003436; RTECS: MD5420000 Solid; Molecular Formula: Au; MW: 196.97 Melting Point: 1063°; Boiling Point: 2808° Density (g/mL): 19.3

  • RTECS: MD5420000
  • Solid


Formula: Au
Boiling Pt: 2808 °C
Melting Pt: 1063 °C
Density: 19.3 g/mL
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Order Now
Specification Test Results
Assay 99.995%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty