Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rhodium acetate

Supplier: BEANTOWN CHEMICAL - BTC

130270-1G 130270-5G
BT130270-1GEA 454 USD InStock
BT130270-1G BT130270-5G
Rhodium acetate
Rhodium acetate
CAS: 42204-14-8; EC No: 255-707-9; MDL No: MFCD01074923 Powder; Molecular Formula: C14H27O20Rh3; MW: 824.07

  • Powder


Formula: C₆H₉O₆Rh
MDL Number: MFCD16556185
CAS Number: 42204-14-8
EINECS: 255-707-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty