Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Manganese(II) carbonate ≥99.95%

Supplier: BEANTOWN CHEMICAL - BTC

130540-100G 130540-25G 130540-5G
BT130540-100GEA 588 USD InStock
BT130540-100G BT130540-25G BT130540-5G
Manganese(II) carbonate ≥99.95%
Manganese(II) carbonate
CAS: 598-62-9; EC No: 209-942-9; MDL No: MFCD00011116 Powder; Linear Formula: MnCO3; MW: 114.95 Melting Point: 350° (decomposes) Density (g/mL): 3.125

  • Powder


Formula: CMnO₃
MW: 114.95 g/mol
Melting Pt: 350 °C (dec.)
Density: 3.125 g/mL
MDL Number: MFCD00011116
CAS Number: 598-62-9
EINECS: 209-942-9
Merck Index: 13,05749
Order Now
Specification Test Results
Assay 99.95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty