130740-100X100
130740-25X25MM
130740-50X50MM
BT130740-100X1EA
1234
USD
InStock
BT130740-100X1
BT130740-25X25
BT130740-50X50
Tantalum ≥99.95% (trace metals basis), foil, Thickness: 1.5 mm
Tantalum
CAS: 7440-25-7; EC No: 231-135-5; MDL No: MFCD00011252
Solid; Molecular Formula: Ta; MW: 180.95
Melting Point: 2996°; Boiling Point: 5425°
Density (g/mL): 16.69