Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lithium salicylate, Technical Grade

Supplier: BEANTOWN CHEMICAL - BTC

130950-100G 130950-25G 130950-500G
BT130950-100GEA 96 USD InStock
BT130950-100G BT130950-25G BT130950-500G
Lithium salicylate, Technical Grade
Lithium salicylate
CAS: 552-38-5; EC No: 209-011-7; MDL No: MFCD00045813 Powder; Molecular Formula: C7H5LiO3; MW: 144.05 Melting Point: 350°

  • Powder


Formula: C₇H₅LiO₃
Melting Pt: >350 °C
MDL Number: MFCD00045813
CAS Number: 552-38-5
EINECS: 209-011-7
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty