131120-25G
131120-5G
BT131120-25GEA
142
USD
InStock
BT131120-25G
BT131120-5G
Diboron trioxide ≥99.999%
Diboron trioxide
CAS: 1303-86-2; EC No: 215-125-8; MDL No: MFCD00011315; RTECS: ED7900000
Powder; Molecular Formula: B2O3 ; MW: 69.62
Melting Point: 45°
Density (g/mL): 2.46
Hygroscopic
- RTECS: ED7900000
- Hygroscopic
- Powder