131215-25CM
131215-5CM
BT131215-25CMEA
637
USD
InStock
BT131215-25CM
BT131215-5CM
Tantalum ≥99.95% (trace metals basis), rods Ø 6.4 mm
Tantalum
CAS: 7440-25-7; EC No: 231-135-5; MDL No: MFCD00011252; RTECS: WW5505000
Solid; Molecular Formula: Ta; MW: 180.95
Melting Point: 2996°; Boiling Point: 5425°
Density (g/mL): 16.69