Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indoline ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

131655-1KG 131655-250G 131655-50G
BT131655-1KGEA 672 USD InStock
BT131655-1KG BT131655-250G BT131655-50G
Indoline ≥99%
Indoline
CAS: 496-15-1; EC No: 207-816-8; MDL No: MFCD00005705; RTECS: NL6906300 Liquid; Molecular Formula: C8H9N; MW: 119.17 Boiling Point: 220-221°; Flash point: 92°C (197°F) Density (g/mL): 1.063; Refractive Index: 1.592

  • RTECS: NL6906300
  • Refractive index: 1.592
  • Liquid


Formula: C₈H₉N
MW: 119.17 g/mol
Boiling Pt: 220…221 °C
Melting Pt: -21
Density: 1.063 g/mL
Flash Pt: 92 °C (197 °F)
MDL Number: MFCD00005705
CAS Number: 496-15-1
EINECS: 207-816-8
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty