Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Methylthio)acetamide

Supplier: BEANTOWN CHEMICAL - BTC

131755-1G 131755-5G
BT131755-1GEA 112 USD InStock
BT131755-1G BT131755-5G
2-(Methylthio)acetamide
2-(Methylthio)acetamide
CAS: 22551-24-2 Solid; Linear Formula: CH3SCH2CONH2; Molecular Formula: C3H7NOS; MW: 105.16 Melting Point: 99-102°; Boiling Point: 259°; Flash point: 110°C (230°F) Density (g/mL): 1.132

  • Solid


Formula: C₃H₇NOS
Boiling Pt: 259 °C
Melting Pt: 99…102 °C
Density: 1.132 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00041321
CAS Number: 22551-24-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty