Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-2-chloro-4-methylpyridine ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

132310-1G 132310-5G
BT132310-1GEA 82 USD InStock
BT132310-1G BT132310-5G
3-Amino-2-chloro-4-methylpyridine ≥97%
3-Amino-2-chloro-4-methylpyridine
CAS: 133627-45-9; MDL No: MFCD00673152 Crystalline; Molecular Formula: C6H7ClN2; MW: 142.59 Melting Point: 65-70°

  • Crystalline


Formula: C₆H₇ClN₂
MW: 142.59
Melting Pt: 65…70 °C
MDL Number: MFCD00673152
CAS Number: 133627-45-9
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty