Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc ferrite

Supplier: BEANTOWN CHEMICAL - BTC

132465-100G 132465-25G
BT132465-100GEA 369 USD InStock
BT132465-100G BT132465-25G
Zinc ferrite
Zinc ferrite
CAS: 12063-19-3; EC No: 235-052-5; MDL No: MFCD00049621 Powder; Linear Formula: ZnFe2O4; MW: 241.08

  • Powder


Formula: Fe₂H₂O₄Zn
MW: 243.082 g/mol
MDL Number: MFCD00049621
CAS Number: 12063-19-3
EINECS: 235-052-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty