Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nickel diethyldithiocarbamate

Supplier: BEANTOWN CHEMICAL - BTC

132715-100G 132715-25G
BT132715-100GEA 105 USD InStock
BT132715-100G BT132715-25G
Nickel diethyldithiocarbamate
Nickel diethyldithiocarbamate
CAS: 14267-17-5; EC No: 238-157-4; MDL No: MFCD00137706 Solid; Molecular Formula: C10H20N2NiS4; MW: 355.23

  • Solid


Formula: C₅H₁₁NS₂
MDL Number: MFCD00137706
CAS Number: 14267-17-5
EINECS: 238-157-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty