132880-10CM
132880-2CM
132880-50CM
BT132880-10CMEA
173
USD
InStock
BT132880-10CM
BT132880-2CM
BT132880-50CM
Silver ≥99.9% (trace metals basis), rods Ø 6.35 mm
Silver
CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397
Solid; Molecular Formula: Ag; MW: 107.87
Melting Point: 960°; Boiling Point: 2212°
Density (g/mL): 10.49