Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichloro-4-methylpyridine

Supplier: BEANTOWN CHEMICAL - BTC

133035-1G 133035-250MG
BT133035-1GEA 318 USD InStock
BT133035-1G BT133035-250MG
3,5-Dichloro-4-methylpyridine
3,5-Dichloro-4-methylpyridine
CAS: 100868-46-0 Solid; Molecular Formula: C6H5Cl2N; MW: 162.02

  • Solid


Formula: C₆H₅Cl₂N
MW: 162.013 g/mol
MDL Number: MFCD09839290
CAS Number: 100868-46-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty