Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Strontium acetate hemihydrate

Supplier: BEANTOWN CHEMICAL - BTC

133135-1KG 133135-250G
BT133135-1KGEA 164 USD InStock
BT133135-1KG BT133135-250G
Strontium acetate hemihydrate
Strontium acetate hemihydrate
CAS: 14692-29-6; EC No: 208-854-8 MDL No: MFCD00198064 Crystalline Linear Formula: (CH3COO)2Sr·0.5H2O MW: 214.72

  • Crystalline


Formula: C₈H₁₄O₉Sr₂
MW: 429.43 g/mol
MDL Number: MFCD00198064
CAS Number: 14692-29-6
EINECS: 208-854-8
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty