Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimetridazole ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

133260-25G 133260-5G
BT133260-25GEA 133 USD InStock
BT133260-25G BT133260-5G
Dimetridazole ≥97%
Dimetridazole
CAS: 551-92-8; EC No: 209-001-2; MDL No: MFCD00047046; RTECS: NI5075000 Solid; Molecular Formula: C5H7N3O2 ; MW: 141.13

  • RTECS: NI5075000
  • Solid


Formula: C₅H₇N₃O₂
MDL Number: MFCD00047046
CAS Number: 551-92-8
EINECS: 209-001-2
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty