Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Silicon dioxide ≥99.5% (trace metals basis)

Supplier: BEANTOWN CHEMICAL - BTC

133480-10KG 133480-2KG
BT133480-10KGEA 131 USD InStock
BT133480-10KG BT133480-2KG
Silicon dioxide ≥99.5% (trace metals basis)
Silicon dioxide
CAS: 7631-86-9; EC No: 238-878-4; MDL No: MFCD00011232; RTECS: VV7330000 -400 Mesh Powder, S.A. 2 m2/g; Linear Formula: SiO2; MW: 60.08 Melting Point: 1610°; Boiling Point: 2230° Density (g/mL): 2.2-2.6; Refractive Index: 1.544

  • RTECS: VV7330000
  • Refractive index: 1.544
  • -400 Mesh Powder, S.A. 2 m²/g


Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1719
Density: 2.2…2.6 g/mL
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 238-878-4
Merck Index: 14,08493
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty