Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-2-Pyrrolidinone-5-carboxylic acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

133885-100G 133885-25G 133885-5G
BT133885-100GEA 848 USD InStock
BT133885-100G BT133885-25G BT133885-5G
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid ≥98%
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid
CAS: 4042-36-8; EC No: 223-735-0; MDL No: MFCD00066212 Crystalline; Molecular Formula: C5H7NO3 ; MW: 129.11 Melting Point: 155-156° Optical Rotation: [α]22/D +10°, c = 1.5 in water

  • Optical rotation: [α]22/D +10°, c = 1.5 in water
  • Crystalline


Formula: C₅H₇NO₃
MW: 129.12 g/mol
Melting Pt: 155…156 °C
MDL Number: MFCD00066212
CAS Number: 4042-36-8
EINECS: 223-735-0
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty