Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dihydrogen hexahydroxyplatinate(IV) ≥99.9%

Supplier: BEANTOWN CHEMICAL - BTC

134145-1G 134145-500MG 134145-5G
BT134145-1GEA 167.93 USD InStock
BT134145-1G BT134145-500MG BT134145-5G
Dihydrogen hexahydroxyplatinate(IV) ≥99.9%
Dihydrogen hexahydroxyplatinate(IV)
CAS: 51850-20-5; EC No: 257-471-2; MDL No: MFCD00058744 Crystalline; Linear Formula: H2Pt(OH)6; MW: 299.17 Hygroscopic

  • Hygroscopic
  • Crystalline


Formula: H₂Pt(OH)₆
MW: 299.14 g/mol
MDL Number: MFCD00058744
CAS Number: 51850-20-5
EINECS: 257-471-2
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty