Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl diethylmalonate ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

134285-100G 134285-25G 134285-500G
BT134285-100GEA 130 USD InStock
BT134285-100G BT134285-25G BT134285-500G
Diethyl diethylmalonate ≥98%
Diethyl diethylmalonate
CAS: 77-25-8; EC No: 201-016-2; MDL No: MFCD00009132 Liquid; Molecular Formula: C11H20O4 ; MW: 216.27 Boiling Point: 228-230°; Flash point: 94°C (201°F) Density (g/mL): 0.99; Refractive Index: 1.423

  • Refractive index: 1.423
  • Liquid


Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
Boiling Pt: 228…230 °C
Density: 0.99 g/mL
Flash Pt: 94 °C (201 °F)
MDL Number: MFCD00009132
CAS Number: 77-25-8
EINECS: 201-016-2
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty