Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl (iodomethyl)phosphonate ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

134365-100G 134365-25G 134365-5G
BT134365-100GEA 1027 USD InStock
BT134365-100G BT134365-25G BT134365-5G
Diethyl (iodomethyl)phosphonate ≥98%
Diethyl (iodomethyl)phosphonate
CAS: 10419-77-9; EC No: 233-895-3; MDL No: MFCD00041386 Liquid; Molecular Formula: C5H12IO3P; MW: 278.03 Boiling Point: 112-115°/1 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.5 Light Sensitive

  • Light Sensitive
  • Liquid


Formula: C₅H₁₂IO₃P
MW: 278.03 g/mol
Boiling Pt: 112…115 °C/1 mmHg
Density: 1.5 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00041386
CAS Number: 10419-77-9
EINECS: 233-895-3
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty