Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydrochlorothiazide ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

134455-100G 134455-25G 134455-5G
BT134455-100GEA 275.48 USD InStock
BT134455-100G BT134455-25G BT134455-5G
Hydrochlorothiazide ≥98%
Hydrochlorothiazide
CAS: 58-93-5; EC No: 200-403-3; MDL No: MFCD00051765; RTECS: DK9100000 Solid; Molecular Formula: C7H8ClN3O4S2; MW: 297.74

  • RTECS: DK9100000
  • Solid


Formula: C₇H₈ClN₃O₄S₂
MDL Number: MFCD00051765
CAS Number: 58-93-5
EINECS: 200-403-3
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty